Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pabpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019402.2 | NP_062275.1 | 2 | 399 | ||
GenBank mRNA | AK032172.1 | - | 2 | 395 | ||
AK157143.1 | - | 2 | 388 | |||
BC049869.1 | - | 2 | 384 | |||
BC055866.1 | - | 2 | 388 | 4218469 |