Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Qdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024236.2 | NP_077198.1 | 3 | 363 | ||
GenBank mRNA | AK011351.1 | - | 3 | 292 | ||
AK078123.1 | - | 3 | 316 | |||
AK159107.1 | - | 3 | 307 | |||
AK161660.1 | - | 3 | 256 | |||
AK166694.1 | - | 3 | 317 | |||
BC002107.1 | - | 3 | 303 | 3487104 |