Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rnf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146025.1 | NP_001139497.1 | 4 | 719 | ||
NM_001146026.1 | NP_001139498.1 | 4 | 644 | |||
NM_001146027.1 | NP_001139499.1 | 4 | 510 | |||
NM_134064.2 | NP_598825.2 | 4 | 655 | |||
GenBank mRNA | AK147349.1 | - | 4 | 645 | ||
AK160090.1 | - | 4 | 506 | |||
AK160115.1 | - | 4 | 506 | |||
AK161651.1 | - | 4 | 632 | |||
AK171943.1 | - | 4 | 501 | |||
AK172257.1 | - | 4 | 501 | |||
BC017630.1 | - | 4 | 448 | 4507289 | ||
BC035548.1 | - | 4 | 470 | 4221284 | ||
CF728349.1 | - | 4 | 574 | |||
CJ147978.1 | - | 4 | 433 |