Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177627.1 | NP_001171098.1 | 2 | 208 | ||
NM_172659.2 | NP_766247.2 | 2 | 208 | |||
GenBank mRNA | AK079650.1 | - | 2 | 152 | ||
AK089246.1 | - | 2 | 200 | |||
AK137441.1 | - | 2 | 174 | |||
AK150104.1 | - | 2 | 161 | |||
AK163445.1 | - | 2 | 170 | |||
AK166312.1 | - | 2 | 166 | |||
AK167904.1 | - | 2 | 170 | |||
AK172411.1 | - | 2 | 152 | |||
BC058210.1 | - | 2 | 132 | |||
BC141168.1 | - | 2 | 139 | 9055780 | ||
BC145480.1 | - | 2 | 139 | |||
BY202464.1 | - | 2 | 209 |