Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cntnap5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172851.2 | NP_766439.2 | 11 | 1743 | ||
GenBank mRNA | AK082656.1 | - | 5 | 801 | ||
BC145618.1 | - | 4 | 508 |