Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178662.3 | NP_848777.1 | 7 | 1031 | ||
GenBank mRNA | AK028298.1 | - | 7 | 1038 | ||
AK038483.1 | - | 7 | 1041 | |||
AK038990.1 | - | 7 | 1032 | |||
AK039411.1 | - | 7 | 1039 | |||
AK086723.1 | - | 7 | 1074 | |||
AK162959.1 | - | 7 | 1039 | |||
AY349150.1 | - | 7 | 996 | |||
BC048903.1 | - | 7 | 1015 | 6491141 |