Search
Search

COX7A1| Gene Symbol: COX7A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001864.3 | NP_001855.1 | 2 | 535 | ||
| GenBank mRNA | AK312091.1 | - | 2 | 374 | ||
| BC002757.2 | - | 2 | 535 | 3631740 | ||
| BT006924.1 | - | 2 | 73 | |||
| CR542120.1 | - | 2 | 73 | |||
| HE647857.1 | - | 2 | 73 | |||
| M83186.1 | - | 2 | 114 | |||