Search
Search

CPB2| Gene Symbol: CPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278541.1 | NP_001265470.1 | 9, 10 | 1034 | ||
| NM_001872.4 | NP_001863.3 | 10, 11 | 1145 | |||
| XM_017020393.1 | XP_016875882.1 | 10, 11 | 1088 | |||
| GenBank mRNA | AB011969.1 | - | 9, 10 | 986 | ||
| AK290829.1 | - | 10, 11 | 1093 | |||
| BC007057.1 | - | 10, 11 | 1102 | 3934520 | ||
| BT006936.1 | - | 10, 11 | 1078 | |||
| M75106.1 | - | 10, 11 | 1097 | |||