Search
Search

GBP1| Gene Symbol: GBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002053.2 | NP_002044.2 | 3 | 438 | ||
| GenBank mRNA | AB208912.1 | - | 3 | 397 | ||
| AK291783.1 | - | 3 | 286 | |||
| AY920436.1 | - | 2 | 222 | |||
| BC002666.2 | - | 3 | 438 | 3606865 | ||
| BT006847.1 | - | 2 | 218 | |||
| M55542.1 | - | 3 | 286 | |||