Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MECP2 SIRNA Assays ›
          Gene Symbol:
          MECP2
          Gene Name:
          methyl-CpG binding protein 2
          Gene Aliases:
          AUTSX3, MRX16, MRX79, MRXS13, MRXSL, PPMX, RS, RTS, RTT
          Chromosome Location:
          Chr.X: 154021800 - 154097731 on Build GRCh38
          UniGene ID:
          Hs.200716
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS106434
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MECP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110792.1 NP_001104262.1 3 551
          NM_001316337.1 NP_001303266.1 5 843
          NM_004992.3 NP_004983.1 4 675
          XM_005274682.4 XP_005274739.1 5 749
          XM_011531166.2 XP_011529468.1 3 562
          XM_017029533.1 XP_016885022.1 7 1175
          GenBank mRNA AF158180.1 - 4 616
          AJ132917.1 - 3 524
          AK289444.1 - 4 650
          AY541280.1 - 3 492
          BC011612.1 - 3 533 3956518
          BX538060.1 - 3 541
          GQ203293.1 - 4 675
          GQ203294.1 - 4 675
          GQ203295.1 - 4 675
          GQ896382.1 - 4 675
          GU479943.1 - 4 675
          GU812285.1 - 3 449
          GU812286.1 - 3 449
          HM020402.1 - 4 675
          HM156732.1 - 4 675
          HM156733.1 - 4 675
          HQ127345.1 - 3 449
          HQ141377.1 - 3 449
          HQ141378.1 - 4 675
          HQ154629.1 - 4 675
          KU178174.1 - 3 449
          L37298.1 - 3 530
          X89430.1 - 2 420
          X99686.1 - 3 523
          Y12643.1 - 3 513

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          behavioral fear response
          response to hypoxia
          startle response
          neurological system process involved in regulation of systemic arterial blood pressure
          regulation of respiratory gaseous exchange by neurological system process
          inositol metabolic process
          mitochondrial electron transport, ubiquinol to cytochrome c
          chromatin silencing
          regulation of gene expression by genetic imprinting
          transcription, DNA-templated
          glutamine metabolic process
          cellular biogenic amine metabolic process
          synapse assembly
          heart development
          respiratory gaseous exchange
          long-term memory
          locomotory behavior
          protein localization
          glucocorticoid metabolic process
          positive regulation of cell proliferation
          adult locomotory behavior
          visual learning
          response to radiation
          pathogenesis
          post-embryonic development
          response to ionizing radiation
          response to lead ion
          dendrite development
          histone methylation
          histone acetylation
          proprioception
          sensory perception of pain
          cerebellum development
          ventricular system development
          negative regulation of histone methylation
          cardiolipin metabolic process
          response to estradiol
          negative regulation of histone acetylation
          social behavior
          neuron maturation
          negative regulation of neuron apoptotic process
          regulation of DNA methylation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          phosphatidylcholine metabolic process
          negative regulation of astrocyte differentiation
          catecholamine secretion
          positive regulation of synapse assembly
          excitatory postsynaptic potential
          long-term synaptic potentiation
          four-way junction DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          mRNA binding
          protein binding
          transcription factor binding
          methyl-CpG binding
          double-stranded methylated DNA binding
          protein domain specific binding
          chromatin DNA binding
          siRNA binding
          histone deacetylase binding
          poly(A) RNA binding
          unmethylated CpG binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zkdp8:80/100.66.75.244:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0