Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MYH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017533.2 | NP_060003.2 | 20 | 2359 | ||
XM_017024676.1 | XP_016880165.1 | 18 | 2503 | |||
GenBank mRNA | AF111783.2 | - | 20 | 2359 |