Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NOLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284388.1 | NP_001271317.1 | 11 | 2061 | ||
NM_001284389.1 | NP_001271318.1 | 11 | 2034 | |||
NM_004741.4 | NP_004732.2 | 11 | 2031 | |||
XM_005270273.1 | XP_005270330.1 | 11 | 2038 | |||
GenBank mRNA | AK056755.1 | - | 10 | 1654 | ||
AK314363.1 | - | 11 | 1818 | |||
AY820769.1 | - | 11 | 1799 | |||
BC001883.1 | - | 11 | 1806 | 3533568 | ||
BC006769.2 | - | 7 | 1316 | 2900024 | ||
CN312242.1 | - | 2 | 217 | |||
D21262.1 | - | 11 | 1822 | |||
Z34289.1 | - | 11 | 1850 |