Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MORF4L2 SIRNA Assays ›
          Gene Symbol:
          MORF4L2
          Gene Name:
          mortality factor 4 like 2
          Gene Aliases:
          MORFL2, MRGX
          Chromosome Location:
          Chr.X: 103675498 - 103688158 on Build GRCh38
          UniGene ID:
          Hs.326387
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS114413
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MORF4L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142418.1 NP_001135890.1 5 854
          NM_001142419.1 NP_001135891.1 4 787
          NM_001142420.1 NP_001135892.1 3 634
          NM_001142421.1 NP_001135893.1 4 736
          NM_001142422.1 NP_001135894.1 5 836
          NM_001142423.1 NP_001135895.1 4 754
          NM_001142424.1 NP_001135896.1 5 803
          NM_001142425.1 NP_001135897.1 4 774
          NM_001142426.1 NP_001135898.1 3 652
          NM_001142427.1 NP_001135899.1 4 756
          NM_001142428.1 NP_001135900.1 5 785
          NM_001142429.1 NP_001135901.1 5 823
          NM_001142430.1 NP_001135902.1 4 701
          NM_001142431.1 NP_001135903.1 4 683
          NM_001142432.1 NP_001135904.1 5 805
          NM_012286.2 NP_036418.1 4 805
          GenBank mRNA AF100620.1 - 4 655
          AF167174.1 - 4 650
          AK056012.1 - 4 642
          AK095298.1 - 2 1567
          AK126115.1 - 6 630
          AK126427.1 - 4 1582
          AK223504.1 - 4 686
          AK302083.1 - 5 586
          BC056899.1 - 5 640 3905425
          BC093013.1 - 5 684 30528536
          BM471743.1 - 4 568
          D14812.1 - 4 655

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          chromatin remodeling
          chromatin silencing
          transcription, DNA-templated
          chromatin modification
          histone deacetylation
          regulation of growth
          histone H4 acetylation
          histone H2A acetylation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of striated muscle cell differentiation
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5688f:80/100.66.73.65:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0