Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DUS4L SIRNA Assays ›
          Gene Symbol:
          DUS4L
          Gene Name:
          dihydrouridine synthase 4 like
          Gene Aliases:
          DUS4, PP35
          Chromosome Location:
          Chr.7: 107563957 - 107578523 on Build GRCh38
          UniGene ID:
          Hs.97627
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS117126
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DUS4L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270419.1 NP_001257348.1 7 866
          NM_181581.2 NP_853559.1 7 1016
          NR_073002.1 7 1116
          NR_073003.1 6 898
          NR_073004.1 6 855
          NR_073005.1 6 748
          XM_005250125.4 XP_005250182.1 6 878
          XM_006715844.2 XP_006715907.1 5 754
          XM_006715845.3 XP_006715908.1 4 665
          XM_011515766.2 XP_011514068.1 7 1010
          XM_011515767.2 XP_011514069.1 6 760
          XM_017011708.1 XP_016867197.1 7 1031
          XM_017011709.1 XP_016867198.1 7 1011
          XM_017011710.1 XP_016867199.1 7 865
          XM_017011711.1 XP_016867200.1 6 793
          XM_017011712.1 XP_016867201.1 6 777
          XM_017011713.1 XP_016867202.1 6 847
          GenBank mRNA AK127802.1 - 3 3940
          AK294095.1 - 6 854
          AK297195.1 - 7 1085
          AK308472.1 - 2 592
          AL523141.3 - 6 868
          AL529385.3 - 6 726
          AW954755.1 - 2 301
          BC111774.1 - 5 1234 40033792
          BQ771928.1 - 2 677
          BX357552.2 - 3 331
          BX395100.2 - 7 825
          CX787147.1 - 5 585
          U62767.1 - 7 814

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA dihydrouridine synthesis
          oxidation-reduction process
          tRNA dihydrouridine synthase activity
          flavin adenine dinucleotide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0