Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PHLDB1 SIRNA Assays ›
          Gene Symbol:
          PHLDB1
          Gene Name:
          pleckstrin homology like domain family B member 1
          Gene Aliases:
          LL5A
          Chromosome Location:
          Chr.11: 118606296 - 118658038 on Build GRCh38
          UniGene ID:
          Hs.504062
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS118329
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PHLDB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144758.2 NP_001138230.1 22, 23 4111
          NM_015157.3 NP_055972.1 23, 24 4334
          XM_005271468.3 XP_005271525.1 23, 24 4925
          XM_005271470.3 XP_005271527.1 22, 23 4880
          XM_005271472.3 XP_005271529.1 22, 23 4784
          XM_005271473.3 XP_005271530.1 21, 22 4739
          XM_005277680.3 XP_005277737.1 26, 27 5366
          XM_006718796.3 XP_006718859.1 26, 27 4555
          XM_006718797.3 XP_006718860.1 25, 26 5225
          XM_006718798.3 XP_006718861.1 24, 25 5054
          XM_006718799.3 XP_006718862.1 23, 24 5009
          XM_006718801.3 XP_006718864.1 23, 24 4913
          XM_006718802.3 XP_006718865.1 22, 23 4868
          XM_011542703.2 XP_011541005.1 26, 27 5369
          XM_011542705.2 XP_011541007.1 25, 26 5324
          XM_011542707.2 XP_011541009.1 25, 26 5240
          XM_011542708.2 XP_011541010.1 26, 27 4558
          XM_011542709.2 XP_011541011.1 26, 27 4868
          XM_011542710.2 XP_011541012.1 25, 26 5198
          XM_011542711.2 XP_011541013.1 23, 24 4275
          XM_011542713.2 XP_011541015.1 23, 24 4269
          XM_017017407.1 XP_016872896.1 25, 26 5237
          XM_017017410.1 XP_016872899.1 24, 25 5096
          GenBank mRNA AB014538.2 - 22, 23 4088
          AB094090.1 - 23, 24 4334

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          regulation of gastrulation
          regulation of epithelial to mesenchymal transition
          regulation of microtubule cytoskeleton organization
          positive regulation of basement membrane assembly involved in embryonic body morphogenesis

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0