Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WDPCP SIRNA Assays ›
          Gene Symbol:
          WDPCP
          Gene Name:
          WD repeat containing planar cell polarity effector
          Gene Aliases:
          BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
          Chromosome Location:
          Chr.2: 63120755 - 63840825 on Build GRCh38
          UniGene ID:
          Hs.414952
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS121553
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WDPCP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042692.2 NP_001036157.1 7, 8 1469
          NM_015910.5 NP_056994.3 13, 14 2284
          NR_122106.1 7, 8 1469
          XM_005264348.3 XP_005264405.1 13, 14 2324
          XM_011532881.2 XP_011531183.1 14, 15 2108
          XM_011532884.2 XP_011531186.1 13, 14 2324
          XM_011532887.2 XP_011531189.1 13, 14 2324
          XM_017004251.1 XP_016859740.1 15, 16 2962
          XM_017004252.1 XP_016859741.1 14, 15 2108
          GenBank mRNA AF131737.1 - 7, 8 1469
          AK022647.1 - 7, 8 1443
          AL833623.1 - 13, 14 2070
          BC093752.1 - 7, 8 1414 7939597
          BC093754.1 - 7, 8 1414 7939599
          BX647132.1 - 7, 8 4797

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          establishment of planar polarity
          kidney development
          auditory receptor cell morphogenesis
          smoothened signaling pathway
          regulation of fibroblast migration
          regulation of embryonic cell shape
          septin cytoskeleton organization
          regulation of protein localization
          cilium assembly
          embryonic digit morphogenesis
          camera-type eye development
          establishment of protein localization
          regulation of focal adhesion assembly
          digestive system development
          palate development
          cilium morphogenesis
          respiratory system development
          cardiovascular system development
          glomerular visceral epithelial cell migration
          regulation of ruffle assembly
          regulation of establishment of cell polarity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hgv68:80/100.66.79.23:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0