Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019074.3 | NP_061947.1 | 4, 5 | 1003 | ||
GenBank mRNA | AB036931.1 | - | 4, 5 | 943 | ||
AB043894.1 | - | 4, 5 | 667 | |||
AF253468.1 | - | 4, 5 | 667 | |||
AF279305.1 | - | 3, 4 | 573 | |||
AK313831.1 | - | 4, 5 | 998 | |||
AY358894.1 | - | 4, 5 | 702 | |||
BC106950.2 | - | 4, 5 | 803 | 40034887 |