Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CMTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099642.1 | NP_001093112.1 | 3 | 2320 | ||
NM_001324374.1 | NP_001311303.1 | 3 | 2251 | |||
NM_001324377.1 | NP_001311306.1 | 3 | 2022 | |||
NM_001324378.1 | NP_001311307.1 | 2 | 2275 | |||
NM_001324379.1 | NP_001311308.1 | 2 | 2337 | |||
NM_018348.5 | NP_060818.4 | 3 | 2464 | |||
XM_011523228.1 | XP_011521530.1 | 3 | 2609 | |||
XM_011523229.1 | XP_011521531.1 | 3 | 2604 | |||
XM_017023444.1 | XP_016878933.1 | 3 | 2807 | |||
GenBank mRNA | AK000289.1 | - | 1 | 853 | ||
AK002033.1 | - | 1 | 2048 | |||
AK023183.1 | - | 1 | 1753 | |||
AK226169.1 | - | 3 | 2215 | |||
AK315056.1 | - | 3 | 2262 | |||
BC034468.1 | - | 3 | 2046 | 4824516 | ||
BC035005.2 | - | 3 | 2251 | 5267637 | ||
BX648660.1 | - | 3 | 2227 |