Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRDM15 SIRNA Assays ›
          Gene Symbol:
          PRDM15
          Gene Name:
          PR domain 15
          Gene Aliases:
          C21orf83, PFM15, ZNF298
          Chromosome Location:
          Chr.21: 41798225 - 41879482 on Build GRCh38
          UniGene ID:
          Hs.473893
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS127280
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRDM15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040424.2 NP_001035514.1 9, 10 1407
          NM_001282934.1 NP_001269863.1 9, 10 1407
          NM_022115.4 NP_071398.3 16, 17 2394
          NR_104257.1 9, 10 1407
          NR_104258.1 9, 10 1347
          NR_104260.1 9, 10 1262
          NR_135464.1 9, 10 1403
          XM_006724039.2 XP_006724102.1 9, 10 1321
          XM_006724040.2 XP_006724103.1 9, 10 1321
          XM_011529674.1 XP_011527976.1 8, 9 1219
          XM_011529675.1 XP_011527977.1 9, 10 1263
          XM_011529676.2 XP_011527978.1 9, 10 3672
          XM_011529677.2 XP_011527979.1 9, 10 1348
          XM_011529678.2 XP_011527980.1 9, 10 1392
          XM_011529679.2 XP_011527981.1 9, 10 1725
          XM_011529681.2 XP_011527983.1 9, 10 2629
          XM_011529683.1 XP_011527985.1 15, 16 2110
          XM_017028425.1 XP_016883914.1 9, 10 1262
          XM_017028426.1 XP_016883915.1 9, 10 3671
          GenBank mRNA AB051812.1 - 9, 10 1398
          AB051813.1 - 9, 10 1398
          AB051814.1 - 9, 10 1398
          AB051815.1 - 9, 10 1338
          AF276513.1 - 16, 17 2394
          AY078498.2 - 16, 17 2394
          BC067102.1 - 9, 10 1262 5925140
          CF138151.1 - 2, 3 304

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          methylation
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding
          methyltransferase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ckh64:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0