Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCDC86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024098.3 | NP_077003.1 | 1 | 857 | ||
GenBank mRNA | AK025974.1 | - | 1 | 743 | ||
AK123775.1 | - | 3 | 530 | |||
AK302329.1 | - | 2 | 647 | |||
BC001378.1 | - | 1 | 708 | 3051044 | ||
DQ501251.1 | - | 1 | 688 |