Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004298.2 | NP_001004298.2 | 3 | 432 | ||
XM_005269491.1 | XP_005269548.1 | 3 | 502 | |||
XM_011539216.1 | XP_011537518.1 | 4 | 1160 | |||
XM_011539217.2 | XP_011537519.1 | 4 | 855 | |||
XM_011539220.2 | XP_011537522.1 | 4 | 400 | |||
XM_011539222.2 | XP_011537524.1 | 4 | 522 | |||
XM_011539223.2 | XP_011537525.1 | 3 | 1210 | |||
XM_011539224.2 | XP_011537526.1 | 3 | 356 | |||
XM_011539226.1 | XP_011537528.1 | 3 | 355 | |||
XM_011539227.2 | XP_011537529.1 | 3 | 688 | |||
XM_011539230.2 | XP_011537532.1 | 1 | 95 | |||
XM_017015634.1 | XP_016871123.1 | 4 | 1160 | |||
XM_017015635.1 | XP_016871124.1 | 4 | 1160 | |||
GenBank mRNA | AK057500.1 | - | 3 | 432 | ||
AK093093.1 | - | 5 | 928 | |||
AK297577.1 | - | 4 | 632 | |||
AK308357.1 | - | 4 | 519 | |||
AK309217.1 | - | 4 | 543 | |||
BC140898.1 | - | 3 | 419 | 9021766 | ||
BC146869.1 | - | 3 | 419 | 9056797 |