Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CPT1C SIRNA Assays ›
Gene Symbol:
CPT1C
Gene Name:
carnitine palmitoyltransferase 1C
Gene Aliases:
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
Chromosome Location:
Chr.19: 49690672 - 49713731 on Build GRCh38
UniGene ID:
Hs.112195
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS133867
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CPT1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136052.2
NP_001129524.1
15
1922
NM_001199752.2
NP_001186681.1
15
2019
NM_001199753.1
NP_001186682.1
15
1955
NM_152359.2
NP_689572.1
14
1886
NR_108072.1
16
2117
XM_005258505.2
XP_005258562.1
15
1992
XM_005258506.4
XP_005258563.1
14
1908
XM_006723009.3
XP_006723072.1
12
1724
XM_011526438.2
XP_011524740.1
15
2048
XM_011526439.2
XP_011524741.1
15
2112
XM_011526440.2
XP_011524742.1
15
1964
XM_017026265.1
XP_016881754.1
15
2011
XM_017026266.1
XP_016881755.1
15
1932
XM_017026267.1
XP_016881756.1
14
1782
XM_017026268.1
XP_016881757.1
13
1625
XM_017026269.1
XP_016881758.1
13
1735
XM_017026270.1
XP_016881759.1
13
1503
XM_017026271.1
XP_016881760.1
13
1592
XM_017026272.1
XP_016881761.1
14
1819
XM_017026273.1
XP_016881762.1
14
1804
GenBank mRNA
AB208884.1
-
9
999
AF357970.1
-
14
1844
AK074389.1
-
7
865
AK096516.1
-
16
2113
AK289713.1
-
15
2011
AK290092.1
-
15
1927
AK299866.1
-
12
1713
AK309087.1
-
14
1845
AL831876.1
-
5
517
BC029104.1
-
15
1886
3872727
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
carnitine metabolic process
carnitine O-palmitoyltransferase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0