Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NLRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317000.1 | NP_001303929.1 | 3 | 1806 | ||
NM_176811.2 | NP_789781.2 | 3 | 1806 | |||
GenBank mRNA | AY154463.1 | - | 3 | 1735 |