Search
Search

SUN5| Gene Symbol: SUN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080675.3 | NP_542406.2 | 11, 12 | 980 | ||
| XM_011528573.1 | XP_011526875.1 | 12, 13 | 1064 | |||
| XM_011528574.1 | XP_011526876.1 | 10, 11 | 920 | |||
| XM_011528575.1 | XP_011526877.1 | 8, 9 | 699 | |||
| GenBank mRNA | AF401350.1 | - | 11, 12 | 980 | ||
| BC026118.2 | - | 11, 12 | 980 | 4824412 | ||
| BC029528.1 | - | 11, 12 | 973 | 5271065 | ||