Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AGBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178563.3 | NP_848658.3 | 7 | 848 | ||
XM_011516122.2 | XP_011514424.1 | 6 | 654 | |||
XM_017012136.1 | XP_016867625.1 | 7 | 2170 | |||
XM_017012137.1 | XP_016867626.1 | 7 | 2452 | |||
XM_017012138.1 | XP_016867627.1 | 6 | 654 | |||
XM_017012139.1 | XP_016867628.1 | 6 | 654 | |||
XM_017012140.1 | XP_016867629.1 | 6 | 654 | |||
XM_017012141.1 | XP_016867630.1 | 6 | 1987 | |||
XM_017012142.1 | XP_016867631.1 | 6 | 654 | |||
XM_017012143.1 | XP_016867632.1 | 6 | 654 | |||
GenBank mRNA | BC030651.2 | - | 7 | 823 | 5267615 |