Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ACYP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320589.1 | NP_001307518.1 | 1 | 446 | ||
NM_001320590.1 | NP_001307519.1 | 1 | 446 | |||
NM_138448.3 | NP_612457.1 | 1 | 446 | |||
XM_017005410.1 | XP_016860899.1 | 1 | 316 | |||
GenBank mRNA | AK307096.1 | - | 1 | 317 | ||
BC012290.1 | - | 1 | 215 | 3864319 | ||
BF739886.1 | - | 1 | 446 | |||
BU857820.1 | - | 1 | 304 | |||
X84195.1 | - | 1 | 195 |