This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
TYROBP
- Gene Name:
-
TYRO protein tyrosine kinase binding protein
- Gene Aliases:
-
DAP12, KARAP, PLOSL
- Chromosome Location:
-
Chr.19: 35904401 - 35908309 on Build GRCh38
- UniGene ID:
-
Hs.515369
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
TYROBP |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_003332.3 |
NP_003323.1 |
2, 3
|
176
|
|
| NM_198125.2 |
NP_937758.1 |
2, 3
|
176
|
|
| GenBank mRNA |
AF019562.1 |
- |
2, 3
|
162
|
|
| AJ010098.1 |
- |
2, 3
|
97
|
|
| AK290385.1 |
- |
2, 3
|
162
|
|
| AY074782.1 |
- |
2, 3
|
96
|
|
| BC011175.1 |
- |
2, 3
|
151
|
4150408 |
| BP302545.1 |
- |
2, 3
|
176
|
|
| BT009851.1 |
- |
2, 3
|
95
|
|
| CR450342.1 |
- |
2, 3
|
95
|
|
| CR542202.1 |
- |
2, 3
|
95
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System