Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_198455.2 | NP_940857.2 | 88 | 13433 | ||
GenBank mRNA | AK123170.1 | - | 4 | 436 | ||
AY927597.1 | - | LIT3590 AY927597 |
2 | 99 | ||
AY927598.1 | - | 3 | 230 |