Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGCC SIRNA Assays ›
Gene Symbol:
RGCC
Gene Name:
regulator of cell cycle
Gene Aliases:
C13orf15, RGC-32, RGC32, bA157L14.2
Chromosome Location:
Chr.13: 41457406 - 41470877 on Build GRCh38
UniGene ID:
Hs.507866
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS147016
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGCC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014059.2
NP_054778.2
2
413
GenBank mRNA
AF036549.1
-
2
197
BC066334.1
-
2
269
5301484
DB451897.1
-
2
413
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of exit from mitosis
negative regulation of endothelial cell proliferation
positive regulation of extracellular matrix constituent secretion
complement activation
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
negative regulation of cell proliferation
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
negative regulation of angiogenesis
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
activation of protein kinase activity
positive regulation of collagen biosynthetic process
negative regulation of blood vessel endothelial cell migration
positive regulation of mitotic nuclear division
positive regulation of transcription from RNA polymerase II promoter
negative regulation of cytokine secretion
positive regulation of cytokine secretion
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of stress fiber assembly
positive regulation of cell cycle arrest
cellular response to hypoxia
mitotic cell cycle arrest
fibroblast activation
negative regulation of fibroblast growth factor production
positive regulation of extracellular matrix assembly
negative regulation of mitotic cell cycle phase transition
negative regulation of cell-cell adhesion mediated by cadherin
positive regulation of endothelial cell apoptotic process
positive regulation of DNA biosynthetic process
protein binding
protein kinase binding
protein kinase activator activity
R-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0