Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: YBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015982.3 | NP_057066.2 | 5 | 744 | ||
XM_017024713.1 | XP_016880202.1 | 6 | 834 | |||
GenBank mRNA | AB208818.1 | - | 2 | 3725 | ||
AF096834.1 | - | 5 | 742 | |||
AK092747.1 | - | 4 | 1361 | |||
BC033800.1 | - | 5 | 707 | 5171696 | ||
BC047760.1 | - | 5 | 708 | 5742121 |