Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138369.2 | NP_612378.1 | 3 | 663 | ||
GenBank mRNA | AK301982.1 | - | 3 | 651 | ||
AY303777.1 | - | 3 | 436 | |||
BC007436.2 | - | 3 | 653 | 2961368 |
Gene Symbol: BOD1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257964.1 | NP_001244893.1 | 1 | 648 | ||
GenBank mRNA | AK127964.1 | - | 1 | 648 | ||
AK301799.1 | - | 2 | 470 | |||
BC021740.1 | - | 1 | 612 | |||
BC035018.1 | - | 1 | 573 |