Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C16orf87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001436.2 | NP_001001436.1 | 1, 2 | 83 | ||
GenBank mRNA | BC056676.1 | - | 1, 2 | 87 | 6711353 | |
BX647945.1 | - | 1, 2 | 83 |