Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CDH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016279.3 | NP_057363.3 | 12 | 2233 | ||
GenBank mRNA | AB035302.1 | - | 12 | 2218 | ||
AK294183.1 | - | 5 | 1080 | |||
AK307779.1 | - | 12 | 2234 | |||
BC107594.1 | - | 12 | 2195 | 40011942 | ||
BC113745.1 | - | 12 | 2195 | 40011941 |