Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ILVBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006844.4 | NP_006835.2 | 6, 7 | 868 | ||
XM_005259717.4 | XP_005259774.1 | 6, 7 | 1015 | |||
XM_011527651.2 | XP_011525953.1 | 5, 6 | 847 | |||
GenBank mRNA | AB209065.1 | - | 3, 4 | 1124 | ||
BC011722.2 | - | 6, 7 | 835 | 4300207 | ||
BC011761.2 | - | 6, 7 | 761 | 3622453 | ||
BC126913.1 | - | 4, 5 | 608 | |||
U61263.1 | - | 6, 7 | 743 |