Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCDC89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152723.2 | NP_689936.1 | 1 | 511 | ||
GenBank mRNA | AK095478.1 | - | 1 | 511 | ||
BC044814.1 | - | 1 | 407 | 5167843 | ||
BI559499.1 | - | 1 | 467 |