Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FUT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002034.2 | NP_002025.2 | 2 | 410 | ||
GenBank mRNA | BC140905.1 | - | 2 | 406 | 9056717 | |
U27329.1 | - | 2 | 410 | |||
U27330.1 | - | 2 | 410 |