Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POLL SIRNA Assays ›
          Gene Symbol:
          POLL
          Gene Name:
          polymerase (DNA) lambda
          Gene Aliases:
          BETAN, POLKAPPA
          Chromosome Location:
          Chr.10: 101578882 - 101588270 on Build GRCh38
          UniGene ID:
          Hs.523230
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS178433
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POLL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001174084.1 NP_001167555.1 9 1896
          NM_001174085.1 NP_001167556.1 9 1557
          NM_001308382.1 NP_001295311.1 6 738
          NM_013274.3 NP_037406.1 9 1833
          NR_033406.1 9 1432
          XM_006717775.1 XP_006717838.1 6 1501
          XM_006717776.1 XP_006717839.1 7 729
          XM_006717777.1 XP_006717840.1 5 1459
          XM_011539650.1 XP_011537952.1 9 1522
          XM_011539651.1 XP_011537953.1 9 1501
          XM_011539652.1 XP_011537954.1 9 1810
          XM_011539653.1 XP_011537955.1 9 1803
          XM_011539654.1 XP_011537956.1 9 1246
          XM_011539655.1 XP_011537957.1 9 1225
          XM_011539656.1 XP_011537958.1 8 1204
          XM_011539657.1 XP_011537959.1 9 1749
          XM_011539659.1 XP_011537961.1 6 1798
          XM_011539660.1 XP_011537962.1 7 1054
          XM_011539662.1 XP_011537964.1 6 1522
          XM_011539663.1 XP_011537965.1 7 750
          XM_011539664.1 XP_011537966.1 5 1480
          XM_011539665.2 XP_011537967.1 6 1068
          XM_011539666.1 XP_011537968.1 6 746
          XM_011539667.1 XP_011537969.1 8 1024
          XM_017016084.1 XP_016871573.1 7 1219
          XM_017016085.1 XP_016871574.1 7 1198
          XM_017016086.1 XP_016871575.1 7 1891
          XM_017016087.1 XP_016871576.1 7 943
          XM_017016088.1 XP_016871577.1 7 1215
          XM_017016089.1 XP_016871578.1 7 1327
          XM_017016090.1 XP_016871579.1 7 1194
          XM_017016091.1 XP_016871580.1 6 1777
          XM_017016092.1 XP_016871581.1 7 1496
          XM_017016093.1 XP_016871582.1 6 1005
          GenBank mRNA AF161019.1 - 8 1462
          AF218027.1 - 3 638
          AF283478.1 - 8 1401
          AJ131890.1 - 9 1772
          AJ844641.1 - 3 339
          AJ844642.1 - 2 207
          AK021600.1 - 9 1504
          AK022476.1 - 9 1410
          AK091106.1 - 2 2016
          AK094956.1 - 2 975
          AK127896.1 - 4 1168
          AK128521.1 - 2 680
          AK292225.1 - 9 1479
          AK293421.1 - 8 1054
          AK293602.1 - 7 729
          AK300862.1 - 6 860
          AY302442.1 - 8 1125
          BC003548.1 - 5 1391 3536366
          BC068529.1 - 9 1785 5267859
          BI906881.1 - 6 698

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-directed DNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          nucleotide-excision repair
          double-strand break repair via nonhomologous end joining
          somatic hypermutation of immunoglobulin genes
          DNA biosynthetic process
          DNA binding
          DNA-directed DNA polymerase activity
          lyase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0