Search
Search

SERPINA4| Gene Symbol: SERPINA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289032.1 | NP_001275961.1 | 4 | 1450 | ||
| NM_001289033.1 | NP_001275962.1 | 4 | 1295 | |||
| NM_006215.3 | NP_006206.2 | 4 | 1135 | |||
| GenBank mRNA | AK309586.1 | - | 4 | 1428 | ||
| AK313196.1 | - | 4 | 1113 | |||
| BC014992.1 | - | 4 | 1109 | 4866432 | ||
| BX248009.1 | - | 4 | 1210 | |||
| BX248760.1 | - | 4 | 1098 | |||
| L19684.1 | - | 3 | 1043 | |||