Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NLRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199138.1 | NP_001186067.1 | 3 | 374 | ||
NM_001199139.1 | NP_001186068.1 | 3 | 257 | |||
NM_001302504.1 | NP_001289433.1 | 3 | 388 | |||
NM_021209.4 | NP_067032.3 | 3 | 388 | |||
XM_017004619.1 | XP_016860108.1 | 3 | 374 | |||
GenBank mRNA | AF376061.1 | - | 4 | 602 | ||
AK095467.1 | - | 3 | 374 | |||
AK314762.1 | - | 3 | 359 | |||
AY027787.1 | - | 3 | 389 | |||
AY027788.1 | - | 3 | 389 | |||
AY027789.1 | - | 3 | 389 | |||
AY027790.1 | - | 3 | 389 | |||
AY032589.1 | - | 2 | 148 | |||
AY035391.1 | - | 3 | 257 | |||
AY358152.1 | - | 3 | 336 | |||
BC031555.1 | - | 3 | 344 | 5179909 | ||
DN998613.1 | - | 3 | 394 |