Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RGS3 SIRNA Assays ›
Gene Symbol:
RGS3
Gene Name:
regulator of G-protein signaling 3
Gene Aliases:
C2PA, RGP3
Chromosome Location:
Chr.9: 113444729 - 113597743 on Build GRCh38
UniGene ID:
Hs.494875
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS184287
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RGS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276260.1
NP_001263189.1
2
987
NM_001282922.1
NP_001269851.1
3
845
NM_001282923.1
NP_001269852.1
18
2455
NM_001322215.1
NP_001309144.1
2
952
NM_130795.3
NP_570613.2
11
1889
NM_144488.5
NP_652759.3
21
2793
XM_006717219.2
XP_006717282.1
20
2639
XM_006717220.1
XP_006717283.1
18
2415
XM_006717222.1
XP_006717285.1
11
1765
XM_006717225.1
XP_006717288.1
2
769
XM_006717226.2
XP_006717289.1
3
1062
XM_006717230.3
XP_006717293.1
2
651
XM_011518894.1
XP_011517196.1
3
954
XM_011518896.1
XP_011517198.1
3
1316
XM_011518897.2
XP_011517199.1
4
1314
XM_011518898.1
XP_011517200.1
4
1038
XM_011518900.1
XP_011517202.1
3
839
XM_011518901.1
XP_011517203.1
3
870
XM_017015003.1
XP_016870492.1
20
2668
XM_017015004.1
XP_016870493.1
17
2163
XM_017015005.1
XP_016870494.1
5
1002
XM_017015006.1
XP_016870495.1
3
1218
GenBank mRNA
AF463495.1
-
11
1889
AF490839.1
-
18
2267
AF490840.1
-
11
1739
AF493927.1
-
1
545
AK125094.1
-
2
1896
AK128127.1
-
21
2791
AK226153.1
-
1
2359
AK301515.1
-
2
849
AK310705.1
-
2
849
AY585192.1
-
1
545
BC042555.1
-
11
1794
5175121
U27655.1
-
3
832
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of GTPase activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0