Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BMP6 SIRNA Assays ›
          Gene Symbol:
          BMP6
          Gene Name:
          bone morphogenetic protein 6
          Gene Aliases:
          VGR, VGR1
          Chromosome Location:
          Chr.6: 7726778 - 7881728 on Build GRCh38
          UniGene ID:
          Hs.285671
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS185916
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BMP6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001718.4 NP_001709.1 2 891
          GenBank mRNA AK300628.1 - 3 851
          BC096084.2 - 3 459
          BC096085.2 - 3 459
          BC099624.1 - 3 459
          BC099625.1 - 3 623
          BC111763.1 - 3 623
          BC112352.1 - 3 623
          BC113018.1 - 3 518
          BC113019.1 - 3 515
          BC128031.1 - 3 415
          M60315.1 - 2 872

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          osteoblast differentiation
          eye development
          kidney development
          positive regulation of endothelial cell proliferation
          endochondral ossification
          type B pancreatic cell development
          cellular iron ion homeostasis
          inflammatory response
          immune response
          positive regulation of pathway-restricted SMAD protein phosphorylation
          response to activity
          positive regulation of bone mineralization
          BMP signaling pathway
          male genitalia development
          positive regulation of lipopolysaccharide-mediated signaling pathway
          response to magnesium ion
          positive regulation of chondrocyte differentiation
          positive regulation of aldosterone biosynthetic process
          response to retinoic acid
          growth
          regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of endothelial cell differentiation
          positive regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of epithelial cell proliferation
          positive regulation of protein secretion
          cartilage development
          response to glucocorticoid
          positive regulation of SMAD protein import into nucleus
          SMAD protein signal transduction
          multicellular organismal iron ion homeostasis
          cellular response to mechanical stimulus
          cellular response to iron ion
          cellular response to BMP stimulus
          positive regulation of DNA-dependent DNA replication
          positive regulation of aldosterone secretion
          cytokine activity
          transforming growth factor beta receptor binding
          growth factor activity
          protein heterodimerization activity
          BMP receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0