Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CAMKV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320147.1 | NP_001307076.1 | 12 | 1855 | ||
NM_024046.4 | NP_076951.2 | 11 | 1948 | |||
GenBank mRNA | AK074856.1 | - | 11 | 1662 | ||
AK074899.1 | - | 12 | 1539 | |||
AK075294.1 | - | 12 | 1550 | |||
AK226073.1 | - | 3 | 699 | |||
AK295588.1 | - | 12 | 1413 | |||
AK297258.1 | - | 11 | 1594 | |||
AK297437.1 | - | 12 | 1495 | |||
AK315525.1 | - | 11 | 1681 | |||
AL136697.1 | - | 10 | 1547 | |||
AL833943.1 | - | 9 | 2006 | |||
BC017363.2 | - | 11 | 1619 | 3940018 | ||
BC019256.1 | - | 11 | 1612 | 3502894 | ||
BX647074.1 | - | 10 | 1790 | |||
CR533508.1 | - | 9 | 1385 |