Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MCTP1 SIRNA Assays ›
Gene Symbol:
MCTP1
Gene Name:
multiple C2 and transmembrane domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 94703690 - 95284575 on Build GRCh38
UniGene ID:
Hs.591248
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS188172
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MCTP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002796.3
NP_001002796.1
13, 14
1745
NM_001297777.1
NP_001284706.1
11, 12
1607
NM_024717.5
NP_078993.4
13, 14
2024
XM_005272082.4
XP_005272139.1
14, 15
2151
XM_005272090.4
XP_005272147.1
11, 12
1386
XM_006714692.3
XP_006714755.1
11, 12
1974
XM_011543649.2
XP_011541951.1
14, 15
2225
XM_011543650.2
XP_011541952.1
12, 13
1202
XM_017009855.1
XP_016865344.1
12, 13
1425
XM_017009856.1
XP_016865345.1
13, 14
1524
XM_017009857.1
XP_016865346.1
12, 13
1271
XM_017009858.1
XP_016865347.1
11, 12
1347
XM_017009859.1
XP_016865348.1
13, 14
1349
XM_017009860.1
XP_016865349.1
12, 13
1446
XM_017009861.1
XP_016865350.1
11, 12
1721
XM_017009862.1
XP_016865351.1
10, 11
1719
XM_017009863.1
XP_016865352.1
10, 11
1347
XM_017009864.1
XP_016865353.1
12, 13
2087
XM_017009865.1
XP_016865354.1
11, 12
1667
GenBank mRNA
AK025997.1
-
2, 3
112
AK057694.1
-
11, 12
1386
AK091330.1
-
6, 7
748
AK297325.1
-
10, 11
1027
AY656715.1
-
13, 14
2024
AY656716.1
-
13, 14
1361
BC030005.2
-
13, 14
1506
3834212
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
calcium-mediated signaling
calcium ion binding
calcium-dependent phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline