Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOCS2 SIRNA Assays ›
          Gene Symbol:
          SOCS2
          Gene Name:
          suppressor of cytokine signaling 2
          Gene Aliases:
          CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2
          Chromosome Location:
          Chr.12: 93569822 - 93626236 on Build GRCh38
          UniGene ID:
          Hs.485572
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS189603
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SOCS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270467.1 NP_001257396.1 3 832
          NM_001270468.1 NP_001257397.1 3 801
          NM_001270469.1 NP_001257398.1 3 799
          NM_001270470.1 NP_001257399.1 3 1071
          NM_001270471.1 NP_001257400.1 2 724
          NM_003877.4 NP_003868.1 3 942
          XM_011538929.1 XP_011537231.1 2 767
          XM_011538935.1 XP_011537237.1 2 767
          XM_011538936.1 XP_011537238.1 2 767
          XM_017020147.1 XP_016875636.1 3 915
          XM_017020148.1 XP_016875637.1 3 805
          XM_017020149.1 XP_016875638.1 3 799
          XM_017020150.1 XP_016875639.1 3 1006
          XM_017020151.1 XP_016875640.1 3 811
          XM_017020152.1 XP_016875641.1 3 840
          XM_017020153.1 XP_016875642.1 3 582
          XM_017020154.1 XP_016875643.1 3 644
          XM_017020155.1 XP_016875644.1 2 767
          XM_017020156.1 XP_016875645.1 3 1057
          GenBank mRNA AB004903.1 - 2 390
          AB006966.1 - 2 352
          AB451318.1 - 2 352
          AF020590.1 - 2 551
          AF037989.1 - 3 669
          AK290546.1 - 3 652
          AK297662.1 - 4 532
          AK313165.1 - 3 779
          BC010399.1 - 2 564 4277425
          BC070039.1 - 2 2457

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          negative regulation of protein kinase activity
          JAK-STAT cascade
          regulation of signal transduction
          positive regulation of signal transduction
          protein ubiquitination
          cytokine-mediated signaling pathway
          response to estradiol
          cellular response to hormone stimulus
          negative regulation of apoptotic process
          negative regulation of JAK-STAT cascade
          negative regulation of insulin receptor signaling pathway
          growth hormone receptor signaling pathway
          protein kinase inhibitor activity
          SH3/SH2 adaptor activity
          growth hormone receptor binding
          insulin-like growth factor receptor binding
          protein binding
          JAK pathway signal transduction adaptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pm9fm:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0