Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD36 SIRNA Assays ›
          Gene Symbol:
          CD36
          Gene Name:
          CD36 molecule
          Gene Aliases:
          BDPLT10, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
          Chromosome Location:
          Chr.7: 80602188 - 80679277 on Build GRCh38
          UniGene ID:
          Hs.120949
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS190374
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000072.3 NP_000063.2 11, 12 1528
          NM_001001547.2 NP_001001547.1 11, 12 1489
          NM_001001548.2 NP_001001548.1 11, 12 1528
          NM_001127443.1 NP_001120915.1 10, 11 1234
          NM_001127444.1 NP_001120916.1 9, 10 1409
          NM_001289908.1 NP_001276837.1 7, 8 1085
          NM_001289909.1 NP_001276838.1 9, 10 1113
          NM_001289911.1 NP_001276840.1 9, 10 1167
          NR_110501.1 9, 10 1105
          XM_005250713.1 XP_005250770.1 11, 12 1470
          XM_005250714.1 XP_005250771.1 11, 12 1437
          XM_005250715.4 XP_005250772.1 11, 12 1595
          GenBank mRNA AK096858.1 - 14, 15 1798
          AK129899.1 - 11, 12 1470
          AK300101.1 - 9, 10 1167
          AK312920.1 - 11, 12 1452
          BC008406.1 - 11, 12 1403 4244251
          HM217020.1 - 9, 10 1211
          HM217021.1 - 9, 10 1211
          HM217022.1 - 9, 10 1211
          HM217023.1 - 8, 9 1031
          HM217024.1 - 9, 10 1211
          HM217025.1 - 8, 9 1023
          HM217026.1 - 7, 8 1085
          KU177961.1 - 9, 10 1114
          M24795.1 - 11, 12 1324
          M98398.1 - 11, 12 1368
          M98399.1 - 11, 12 1368
          S67532.1 - 10, 11 1250

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          positive regulation of cell-matrix adhesion
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          platelet degranulation
          MyD88-dependent toll-like receptor signaling pathway
          lipid metabolic process
          receptor-mediated endocytosis
          phagocytosis, recognition
          immune response
          cell adhesion
          cell surface receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          nitric oxide mediated signal transduction
          blood coagulation
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cholesterol storage
          lipid storage
          cGMP-mediated signaling
          positive regulation of blood coagulation
          intestinal cholesterol absorption
          cholesterol transport
          receptor internalization
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          response to lipid
          triglyceride transport
          plasma lipoprotein particle clearance
          low-density lipoprotein particle clearance
          response to stilbenoid
          toll-like receptor TLR6:TLR2 signaling pathway
          lipoprotein transport
          negative regulation of transcription factor import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          apoptotic cell clearance
          negative regulation of growth of symbiont in host
          long-chain fatty acid import
          interleukin-1 beta secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-positive bacterium
          intestinal absorption
          sensory perception of taste
          low-density lipoprotein particle mediated signaling
          positive regulation of phagocytosis, engulfment
          positive regulation of macrophage cytokine production
          positive regulation of ERK1 and ERK2 cascade
          cholesterol import
          response to fatty acid
          response to linoleic acid
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          cellular response to low-density lipoprotein particle stimulus
          cellular response to hydroperoxide
          cellular response to diacyl bacterial lipopeptide
          positive regulation of NLRP3 inflammasome complex assembly
          amyloid fibril formation
          regulation of removal of superoxide radicals
          positive regulation of blood microparticle formation
          positive regulation of reactive oxygen species metabolic process
          regulation of energy homeostasis
          low-density lipoprotein receptor activity
          protein binding
          high-density lipoprotein particle binding
          lipid binding
          low-density lipoprotein particle binding
          transforming growth factor beta binding
          thrombospondin receptor activity
          lipoteichoic acid receptor activity
          lipoprotein particle binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jww4n:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0