Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Elk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282967.1 | NP_001269896.1 | 2 | 469 | ||
NM_013508.2 | NP_038536.2 | 2 | 469 | |||
GenBank mRNA | AK052546.1 | - | 2 | 146 | ||
AK088129.1 | - | 2 | 284 | |||
AK133847.1 | - | 2 | 165 | |||
BC005686.1 | - | 2 | 451 | 3155850 | ||
BC053402.1 | - | 2 | 220 | 30060743 | ||
BC055735.1 | - | 2 | 242 | 6808738 | ||
D26065.1 | - | 1 | 29 | |||
FN434119.1 | - | 1 | 35 | |||
FN434120.1 | - | 1 | 35 | |||
L19953.1 | - | 2 | 360 | |||
S82864.1 | - | 2 | 152 | |||
Z32815.1 | - | 2 | 353 |