Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025559.3 | NP_001020730.1 | 12 | 1477 | ||
NM_001025560.2 | NP_001020731.1 | 12 | 1591 | |||
NM_001277326.1 | NP_001264255.1 | 12 | 1572 | |||
NM_001277327.1 | NP_001264256.1 | 11 | 1632 | |||
NM_001277328.1 | NP_001264257.1 | 10 | 1337 | |||
NM_011445.4 | NP_035575.1 | 13 | 1625 | |||
GenBank mRNA | AK029696.1 | - | 10 | 1265 | ||
AK030713.1 | - | 12 | 1592 | |||
AK048143.1 | - | 12 | 1478 | |||
AK084290.1 | - | 12 | 1633 | |||
AK132409.1 | - | 12 | 1538 | |||
BC067407.1 | - | 11 | 1629 | 30094413 | ||
D61689.1 | - | 10 | 1333 | |||
U32614.1 | - | 13 | 1629 |