Search
Search

Serpine2| Gene Symbol: Serpine2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009255.4 | NP_033281.1 | 8 | 1274 | ||
| GenBank mRNA | AK045954.1 | - | 8 | 1246 | ||
| AK047856.1 | - | 8 | 1246 | |||
| AK051332.1 | - | 8 | 1246 | |||
| AK159687.1 | - | 8 | 1246 | |||
| BC010675.1 | - | 8 | 1224 | 3595407 | ||
| CT010311.1 | - | 7 | 1097 | |||
| X70296.1 | - | 8 | 1245 | |||