Search
Search

Gprc6a| Gene Symbol: Gprc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153071.1 | NP_694711.1 | 6 | 1897 | ||
| GenBank mRNA | AY101365.1 | - | 6 | 1897 | ||
| AY735399.1 | - | 6 | 1870 | |||
| AY735400.1 | - | 6 | 1345 | |||
| AY735401.1 | - | 5 | 1657 | |||
| BC108998.2 | - | 6 | 1897 | 40048019 | ||
Set Membership: |
|