Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cul3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016716.4 | NP_057925.1 | 8 | 1205 | ||
GenBank mRNA | AF129738.1 | - | 8 | 1095 | ||
AK129174.1 | - | 9 | 1593 | |||
BC027304.1 | - | 8 | 1230 | 3981170 |